To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Species:Human
- Size:100 µg
- Protein/peptide name:Glucagon like Peptide 2
- Purity:>90%
- Cat. no.:89158-724
- Supplier No.:GP1439-0100
Spécifications
A propos de cet article
Glucagon-like peptide 2 is a 33- or 34-amino acid peptide located at the C-terminus of, and cleaved from, pro-glucagon by prohormone convertase 3 (PC3). GLP-2 is a potent intestinotropic growth factor, the action of which is transduced by G protein-coupled membrane-bound receptors (GLP2-R) found in the gut. GLP-2Rs are also found in the rodent CNS. The main actions of GLP-2 appear to be crypt cell proliferation, inhibition of enterocyte apoptosis, enhancement of nutrient adsorption and certain reparative effects. Applications: antiserum adsorption control; standard or tracer for immunoassay.