ContactUsLinkComponent
Calcineurin Cellular Activity Assay Kit, Enzo Life Sciences
Calcineurin Cellular Activity Assay Kit, Enzo Life Sciences
# de catalogue 89156-556
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Calcineurin Cellular Activity Assay Kit 1 Kit
Calcineurin Cellular Activity Assay Kit, Enzo Life Sciences
# de catalogue 89156-556
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    96 Wells
  • Application:
    Colorimetric Detection
  • Cat. no.:
    89156-556
  • Supplier No.:
    AK816-0001
Spécifications
A propos de cet article
A complete colorimetric assay kit for measuring cellular calcineurin (PP2B) phosphatase activity
The RII phosphopeptide substrate, supplied with this kit is the most efficient and selective peptide known for calcineurin. The detection of free-phosphate released is based on the classic malachite green assay.
It employs a convenient 96-well plate format with all reagents necessary for measuring calcineurin (PP2B) phosphatase activity in tissue/cellular extracts PLUS human recombinant calcineurin is included as a positive control.
Delivery: Kit includes Calcineurin enzyme, Calmodulin (human, recombinant), Substrate (RII phosphopeptide), 2x assay buffer, EGTA Buffer, Lysis Buffer, Protease inhibitor cocktail, Biomol Green™ Reagent, Phosphate standard, Okadaic acid, Desalting column and resin, Half Volume microplate.
Customers Who Bought This Also Bought