ContactUsLinkComponent
Neutrophil elastase colorimetric drug discovery kit, Enzo Life Sciences
Neutrophil elastase colorimetric drug discovery kit, Enzo Life Sciences
# de catalogue 89156-496
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Neutrophil Elastase Colorimetric Drug Discovery Kit 1 Kit
Neutrophil elastase colorimetric drug discovery kit, Enzo Life Sciences
# de catalogue 89156-496
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    1 Kit
  • Cat. no.:
    89156-496
  • Supplier No.:
    AK497-0001
Spécifications
A propos de cet article
It is a complete assay system designed to screen inhibitors of neutrophil elastase, providing enough material to perform at least 96 assays. Neutrophil elastase, also known as leukocyte elastase (LE), is a serine protease found in polymorphonuclear neutrophils (PMNs) that functions in inflammation. It can cleave a variety of proteins, including fibrous elastin, collagen, clotting factors, TNF-α, and α1-antichymotrypsin. It has been shown to be taken up by tumor cells and accelerate tumor growth through its degradation of insulin receptor substrate-1 (IRS-1). Because of its potential roles in cancer and inflammatory diseases such as emphysema, cystic fibrosis, and atherosclerosis, it is a target for drug discovery. The kit uses the chromogenic substrate MeOSuc-Ala-Ala-Pro-Val-pNA; cleavage of p-nitroaniline (pNA) from the substrate increases absorbance at 405nm. A neutrophil elastase inhibitor, elastatinal, is included for use as a control.
Ordering information: Quantity: 96 assays Contents: Neutrophil elastase (human), (purified) (Prod. No. BML-SE284-9090)(30mU; One U=1μmole/min of MeOSuc-AAPV-pNA at 25°C, pH 8.0. Purity ≥95% (SDS-Page))Storage: -70°C; avoid freeze/thaw cycles.pNA substrate (MeOSuc-AAPV-pNA) (Prod. No. BML-P213-9090)(60μl; 20mM in DMSO)Storage: -70°CpNA calibration standard (p-nitroaniline) (Prod. No. BML-KI106)(1ml; 50μM in assay buffer)Storage: -70°CElastatinal (inhibitor) (Prod. No. BML-PI103-9090)(10μl; 20mM in DMSO)Storage: -70°CNeutrophil elastase assay buffer (Prod. No. BML-KI581)(20ml; 100mM HEPES, pH 7.25, 500mM NaCl, 0.05% Tween-20; liquid in a screw-cap plastic bottle)Storage: -20°C or -70°C96-well microplate (Prod. No. BML-KI101)Storage: Room temperature
Customers Who Bought This Also Bought