ContactUsLinkComponent
Cytochrome C Releasing Apoptosis Assay Kit, Enzo Life Sciences
Cytochrome C Releasing Apoptosis Assay Kit, Enzo Life Sciences
# de catalogue 89156-084
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Cytochrome C Releasing Apoptosis Assay Kit 1 Kit
Cytochrome C Releasing Apoptosis Assay Kit, Enzo Life Sciences
# de catalogue 89156-084
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    1 Kit
  • Storage conditions:
    –20 °C (Long Term)
  • Cat. no.:
    89156-084
  • Supplier No.:
    850-277-KI01
Spécifications
A propos de cet article
For the detection of Cytochrome c translocation from mitochondria into cytosol during apoptosis.
The kit provides an easy-to-use procedure and reagents for separating a mitochondria-enriched fraction from cytosol. Cytochrome c released from mitochondria into cytosol is determined by Western blot using the cytochrome c antibody provided in the kit. The procedure is simple, straightforward and requires no ultracentrifugation.
Delivery: Kit includes Mitochondria Extraction Buffer (10 mL), 5X Cytosol Extraction Buffer (20 mL), 1 M DTT (110 µL), 500X Protease Inhibitor Cocktail (1 vial), 0.2 mg/mL Cytochrome c Antibody (0.5 mL).
Customers Who Bought This Also Bought