Order Entry
ContactUsLinkComponent
Caspase inhibitor set lV, Enzo Life Sciences
Caspase inhibitor set lV, Enzo Life Sciences
# de catalogue 89155-998
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Caspase inhibitor set lV, Enzo Life Sciences
# de catalogue 89155-998
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Size:
    1 Set
  • Formulation:
    Liquid. In DMSO.
  • Cat. no.:
    89155-998
  • Supplier No.:
    850-239-KI01

Spécifications

A propos de cet article

Potent, cell permeable synthetic peptides that irreversibly inhibit activities of each of the caspase-family proteases. YVAD, VDVAD, DEVD, LEVD, WEHD, VEID, IETD, LEHD, and AEVD are the recognition sequences for caspase-1, -2, -3, -4, -5, -6, -8, -9, and -10. VAD is a general caspase inhibitor that inhibits all known caspases. Z-FA-FMK is a negative control for FMK inhibitors.

Ordering information: Contents: Contains 25µl (2mM) each of the following inhibitors:
- Caspase-1 Inhibitor, Z-YVAD-FMK, yellow cap
- Caspase-2 Inhibitor, Z-VDVAD-FMK, orange cap
- Caspase-3 Inhibitor, Z-DEVD-FMK, red cap
- Caspase-4 Inhibitor, Z-LEVD-FMK, orange cap
- Caspase-5 Inhibitor, Z-WEHD-FMK, clear cap
- Caspase-6 Inhibitor, Z-VEID-FMK, violet cap
- Caspase-8 Inhibitor, Z-IETD-FMK, green cap
- Caspase-9 Inhibitor, Z-LEHD-FMK, blue cap
- Caspase-10 Inhibitor, Z-AEVD-FMK, amber cap
- Caspase-13 Inhibitor, Z-LEED-FMK, yellow cap
- Caspase-Family Inhibitor, Z-VAD-FMK, amber cap
- FMK Negative Control, Z-FA-FMK, clear cap
Customers Who Bought This Also Bought