ContactUsLinkComponent
Apoptosis inducer set, Enzo Life Sciences
Apoptosis inducer set, Enzo Life Sciences
# de catalogue 89155-990
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Apoptosis inducer set, Enzo Life Sciences
# de catalogue 89155-990
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Format:
    Liquids in DMSO
  • Description:
    Apoptosis inducer set
  • Size:
    1 Set
  • Cat. no.:
    89155-990
  • Supplier No.:
    850-235-KI01
Spécifications
A propos de cet article
The set contains 5 ready-to-use reagents that induce apoptosis through different mechanisms. Actinomycin D, an antineoplastic antibiotic, inhibits RNA synthesis. Camptothecin, an inhibitor of nuclear topoisomerase, induces apoptosis in many types of cells. Cycloheximide, an active antibiotic against many yeast and fungi, inhibits protein synthesis. Dexamethasone, an active and highly stable glucocorticoid, probably induces apoptosis by binding and activating the intracellular glucocorticoid receptor. Etoposide, a derivative of podophyllotoxin, inhibits topoisomerase activity.
Ordering information: Contents: Contains 5 ready-to-use reagents:
- Actinomycin D, 50µl (10mM), violet cap
- Camptothecin, 1ml (2mM), red cap
- Cycloheximide, 1ml (100mM), green cap
- Dexamethasone, 1ml (10mM), blue cap
- Etoposide, 100µl (100mM), yellow cap
Customers Who Bought This Also Bought