ContactUsLinkComponent
APO-BRDU™ kit, TUNEL Assay
APO-BRDU™ kit, TUNEL Assay
# de catalogue 89155-906
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Apo-Brdu* Kit 1 Kit
APO-BRDU™ kit, TUNEL Assay
# de catalogue 89155-906
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    1 Kit
  • Cat. no.:
    89155-906
  • Supplier No.:
    850-042-KI01
Spécifications
A propos de cet article
Kit for measuring apoptosis with high sensitivity by Dual Color Flow Cytometry in mammalian cells. Br-dUTP is more readily incorporated into the genome of apoptotic cells than are larger ligands like fluorescein, biotin, etc. This greater incorporation coupled with the use of a fluorescein labelled anti-BrdU monoclonal antibody (Prod. No. ALX-804-197L) gives rise to a stronger signal in flow cytometry.
Ordering information: Quantity: 50 assays. Contents: Contains all the reagents necessary, including positive and negative control cells.
Customers Who Bought This Also Bought