To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:BCL2-like 13
- Clonality:Monoclonal
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:BCL2L13
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid. In PBS containing 0.02% sodium azide.
- Antigen synonyms:Bcl2-L-13|BCL-RAMBO|MIL1
- Concentration:1mg/ml
- Purification:≥95% (SDS-PAGE)
- Cat. no.:89155-052
- Supplier No.:804-821-C100
Spécifications
A propos de cet article
Host: Mouse, Isotype: IgG1
- For Immunocytochemistry, Immunoprecipitation, Western Blot
Recognizes the C-terminus of human Bcl-rambo (aa 224-459). Detects endogenous protein by ICC, IP and WB.
Type: Primary
Antigen: BCL2L13
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human