ContactUsLinkComponent
ZnAF-2 DA, Enzo Life Sciences
ZnAF-2 DA, Enzo Life Sciences
# de catalogue 89152-916
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Znaf-2 Da 1 Mg
ZnAF-2 DA, Enzo Life Sciences
# de catalogue 89152-916
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Concentration:
    ~5mM in 0.3 ml DMSO
  • Description:
    ZnAF-2 DA
  • Size:
    1 mg
  • Purity:
    ≥98% (HPLC)
  • Cat. no.:
    89152-916
  • Supplier No.:
    620-076-M001
Spécifications
A propos de cet article
Fluorescent probe for Zn2+
Cell permeable derivative of ZnAF-2 (Prod. No. ALX-620-072). Fluorescent reagent (Ex(max): 492nm; Em(max): 514nm) for the detection of low concentration of zinc ion due to its strong affinity to zinc ion (dissociation constant: 2.7nM). The sample zinc ion can be specifically detected. Low background fluorescense supersensitizes the visualization for in vivo sample zinc ion.
Cell permeable.
Customers Who Bought This Also Bought