Mini Environmental Quality Meter, Sper Scientific
# de catalogue 97036-036
Fournisseur: Sper Scientific
Spécifications
- Description:Mini Environmental Quality Meter
- Dimensions:150 × 65 × 30 mm (6 × 2.5 × 1")
- Cat. no.:97036-036
- Supplier No.:850070
Spécifications
A propos de cet article
For environmental testing anywhere
Combines an anemometer, hygrometer, thermometer, and a light meter into a single compact unit weighing only 5½ oz (160 g). Features dual display with low battery and over range indications, touch-tone buttons, min/max, auto power off and hold functions.
Ordering information: 1-year warranty. Comes ready to use with soft carrying case, wrist strap, instructions, and a 9V battery.
Customers Who Bought This Also Bought