Environmental Quality Meter with Sound, Sper Scientific
Fournisseur: Sper Scientific
Certificats
A propos de cet article
Six meters in one
Combines an anemometer, hygrometer, thermometer, sound and light meter into a single hand-held unit. Also reads any type-K thermocouple probe. Results are displayed on a large dual LCD with low battery and out-of-range indications. Features touch-tone buttons, min/max, auto power off and hold functions.
Certifications: N.I.S.T. traceable certificate of calibration available (for Sound only; Certified model only)
BodyTextAccessories: Simply place the calibrator on to the digital sound meter’s microphone, turn on the calibrator, and adjust the decibel meter's reading to either 94 or 114 dB. Operates at a frequency of 1KHz ±4% and with an accuracy of ±0.5 dB. Built in battery check. Easily portable for field use, it comes with a carrying case, adaptors for 0.5 and 1" microphones, instructions and a 9-volt battery. It conforms to ANSI S1.4-1984 and IEC942 1988 Class 2 standards for acoustical calibrators. 5-year warranty.
Ordering information: 5-year warranty. Comes ready to use in a soft carrying case with RS232 computer port, instructions, 6 AAA batteries, easel back, and a tripod screw.
Spécifications
- Catalog No:
- 97036-032
- 97036-034
- Dimensions:
- 255 × 75 × 35 mm (10 × 2.75 × 1.125")
- 255 × 75 × 35 mm (10 × 2.75 × 1.125")
- Weight:
- 330 g (12 oz.)
- 330 g (12 oz.)