Upchurch Scientific® Frit-In-A-Ferrule™, IDEX Health & Science
Fournisseur: Upchurch Scientific
Certificats
A propos de cet article
Upchurch Scientific® Frit-In-A-Ferrule™ seal and filter simultaneously by incorporating a frit into the body of a flat-bottom ferrule.
Use a Frit-In-A-Ferrule™ to filter at any point in your system where 1/16" or 1/8" OD tubing is used in a flat-bottom 1/4-28, M6 or 5/16-24 connection. This product line is available in both Flangeless and Super Flangeless™ versions.
Use a Frit-In-A-Ferrule™ to filter at any point in your system where ¹/₁₆ or ¹/₈" Ext.Ø tubing is used in a flat-bottom 1/4-28, M6 or 5/16-24 connection. This product line is available in both Flangeless and Super Flangeless™ versions.
This simple yet innovative design allows one to eliminate traditional inline filters and reduce the number of additional connections in a system.
Certifications: These products are RoHS certified.
Ordering information: The Super Flangeless™ versions for ¹/₈" Ext.Ø tubing can not be used in M6 ports.
Customers Who Bought This Also Bought