Nichipet Premium Digital Micropipettors, Nichiryo America
Fournisseur: Nichiryo America
Certificats
A propos de cet article
Innovation in durability, strength, stability, and comfort results in the latest model of the Nichipet series.
- Easy-calibration function provides simple lab calibration
- Enlarged push button makes it easy to depress
Seal ring for the Premium series is tested dispensing/aspirating 600000 times (2 to 1000 μl). Nozzle top is highly abrasion tolerant (2 to 1000 μl). Non-corrosive ceramic is used for the plunger (100 μl to 10 ml).
A Hyper Blower system, with longer second-push stroke, improves dispensing efficiency (2, 10,and 20 μl). Hand temperature does not permeate through the body of the pipettor, which works to avoid affecting volume measurement.
The new, round shape of the Premium series pipettors mitigates fatigue and stress from heavy workloads.
Customers Who Bought This Also Bought