Nichipet EX Plus Digital Micropipettors, Nichiryo America
Fournisseur: Nichiryo America
Certificats
A propos de cet article
Chemical-resistant model of the Nichipet series provides advanced durability against organic solvents dispensing.
- Feature a perfluoro rubber O-Ring with low cubical expansion in the airtight parts
- All models have plungers constructed of materials resistant to organic solvents
- Alloy spring is also included to avoid metal corrosion by organic solvents
- Hyper Blower System, with longer second-push stroke, improves dispensing efficiency
- Enhanced UV resistance enables improved performance in UV hoods
- Easy-calibration function provides simple lab calibration
Ceramic plungers are used for volume models larger than 100 μl. The one-touch locking mechanism makes it easy to lock the sample volume. Fully autoclavable (121 ℃ for 20 minutes).
Customers Who Bought This Also Bought