ContactUsLinkComponent
Ward's® AP Chemistry Investigation 12: Calorimetry: How Does a Hand Warmer Work?
Ward's® AP Chemistry Investigation 12: Calorimetry: How Does a Hand Warmer Work?
# de catalogue 470303-018
Fournisseur:  Ward's Science
Ap chem 12: calorimetry
Ward's® AP Chemistry Investigation 12: Calorimetry: How Does a Hand Warmer Work?
# de catalogue 470303-018
Fournisseur:  Ward's Science
Spécifications
  • Description:
    Ward's® AP Chemistry Investigation 12: Calorimetry
  • No. of Lab Groups:
    15
  • Cat. no.:
    470303-018
  • Includes:
    Teacher's Guide and Student Copymaster
  • Supplier No.:
    470303-018
Spécifications
A propos de cet article
Meets AP Science Practices 1, 2, 4, 5, 6, and 7
  • Addresses Big Idea 5
  • Materials provided for 30 students working in pairs
  • Investigation includes updated literature aligned with new AP Chemistry curriculum standards
In this lab, thermochemistry will be explored, as students conceptualize the chemistry of a hand warmer that is safe, cost effective, eco-friendly, and performs well. Students will use calorimetry to study the heat flow and enthalpy of dissolution of various aqueous solutions, and will therefore be required to set up a calorimeter and calculate changes in temperature, heat transfer, and the enthalpy of dissolution. The investigation begins as students study the heat flow of an aqueous solution of the ionic compound magnesium sulfate. Students will then study the heat flow of a variety of other ionic compounds and will determine the enthalpy of dissolution for these processes. The information learned from these studies will ultimately be used to determine which ionic solid would be best to use in the design of a hand warmer. Includes anhydrous calcium chloride, sodium chloride, anhydrous sodium carbonate, anhydrous magnesium sulfate, ammonium chloride, and lithium chloride. Requires a thermometer, analytical balance, magnetic stirrer, magnetic stir rod, coffee cups with lids, and hotplates, available separately. Ward's® DataHub not required to complete activity.
AP and Advanced Placement Programs are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.
Ordering information: Activity includes Anhydrous Calcium Chloride, Anhydrous Sodium Carbonate, Lithium Chloride, Ammonium Chloride, Sodium Chloride, and Anhydrous Magnesium Sulfate. Materials required but not provided are DI Water, Thermometers, Analytical Balances, Graduated Cylinders, Scoops, Weighing Boats, Polystyrene Cups with Lids, Magnetic Stirrers with Stir Bars, Hot Plates, Beakers, Ring Stands/Clamps (optional), Gloves, Safety Goggles, and Lab Aprons.
Customers Who Bought This Also Bought