ContactUsLinkComponent
Ward's® AP Chemistry Investigation 4: Titrations: How Acidic are the Beverages we Drink?
Ward's® AP Chemistry Investigation 4: Titrations: How Acidic are the Beverages we Drink?
# de catalogue 470303-032
Fournisseur:  Ward's Science
Ap chem 4: titration
Ward's® AP Chemistry Investigation 4: Titrations: How Acidic are the Beverages we Drink?
# de catalogue 470303-032
Fournisseur:  Ward's Science
Spécifications
  • Description:
    Ward's® AP Chemistry Investigation 4: Titrations
  • No. of Lab Groups:
    15
  • Cat. no.:
    470303-032
  • Includes:
    Teacher's Guide and Student Copymaster
  • Supplier No.:
    470303-032
Spécifications
A propos de cet article
Meets AP Science Practices 1, 2, 3, 4, 5, and 6
  • Addresses Big Ideas 1 & 3
  • Materials provided for 30 students working in pairs
  • Investigation includes updated literature aligned with new AP Chemistry curriculum standards
In this investigation, students will explore acid-base reactions using the analytical method of titration. In titration studies, an indicator is often used in order to determine the end point of the reaction via color change. In this activity, students will determine how the volume of the titrant added relates to the morality of the analyte. Students will apply this information as they decide which indicator to use in order to determine the pH of common juices and drinks. Includes orange juice, hydrochloric acid, and sodium hydroxide. requires 50 mL burettes and pH meter, available separately.
AP and Advanced Placement Programs are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.
Includes:
Orange Juice
Coffee
Tea
Methyl Red Solution
Bromothymol Blue Solution
Phenolphthalein Solution
Acetic Acid
Sodium Hydroxide
Hydrochloric Acid
Required but Not Provided: Beakers, Burette and Clamp, Erlenmeyer Flasks, Graduated Cylinders, Volumetric Pipettes, Ring Stands, pH Meters, DI water, Lab Notebook, Gloves, Safety Goggles, Lab Aprons, Additional Fruit Juices/soda (optional).
Customers Who Bought This Also Bought