FA-Leu-Gly-Pro-Ala-OH
Fournisseur: Bachem Americas
Certificats
A propos de cet article
The FA-peptides M-1400 and M-1410 are common ACE substrates and thus filed in the corresponding product family. For our 3-(2-furyl)acryloyl-amino acids (FA-amino acids) and the labeling reagent FA-OSu (Q-1245), please see the Amino Acids section.
Substrate for a versatile, convenient, continuously recording spectrophotometric assay of collagenase.
Customers Who Bought This Also Bought