Ac-Lys(Ac)-D-Ala-D-Ala-OH
Fournisseur: Bachem Americas
Certificats
A propos de cet article
See also F-3260.
Diacetyl-Lys-D-Ala-D-Ala (DALAA) is a substrate for penicillin-sensitive D-alanine carboxypeptidases (DD-carboxypeptidases).
Customers Who Bought This Also Bought