Suc-Phe-Ala-Ala-Phe-pNA
Fournisseur: Bachem Americas
Certificats
A propos de cet article
This enzyme substrate was used to test the concept of substrate-assisted catalysis with an engineered bacterial serine endopeptidase, subtilisin. Suc-FAAF-pNA has been employed for assaying subtilisins from psychrophilic bacteria and myroilysin, a metalloproteinase from the deep sea bacterium Myroides profundi.
Customers Who Bought This Also Bought