H-Glu-Gly-Arg-pNA Acetate
# de catalogue L-1455.0050BA
Fournisseur: Bachem Americas
Numéro CAS:
Spécifications
- CAS No.:67615-71-8
- Size:50 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD02259650
- Protein/peptide name:H-Glu-Gly-Arg-pNA Acetate
- Molecular weight:480.48
- Sequence:H-Glu-Gly-Arg-pNA
- Molecular formula:C₁₉H₂₈N₈O₇
- Cat. no.:L-1455.0050BA
- Supplier No.:L-1455.0050
Spécifications
A propos de cet article
EGR-pNA, a sensitive substrate for the u-plasminogen activator (uPA). Also cleaved by trypsin and matriptase.
Customers Who Bought This Also Bought