H-D-Leu-Thr-Arg-pNA Acetate
# de catalogue L-1380.0050BA
Fournisseur: Bachem Americas
Numéro CAS:
Spécifications
- CAS No.:122630-72-2
- Size:50 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD02259649
- Protein/peptide name:H-D-Leu-Thr-Arg-pNA Acetate
- Molecular weight:508.58
- Sequence:H-D-Leu-Thr-Arg-pNA
- Molecular formula:C₂₂H₃₆N₈O₆
- Cat. no.:L-1380.0050BA
- Supplier No.:L-1380.0050
Spécifications
A propos de cet article
D-LTR-pNA, chromogenic substrate cleaved by recombinant human tryptases ε (PRSS22) and γ (transmembrane tryptase, TMT).
Customers Who Bought This Also Bought