H-D-Ile-Phe-Lys-pNA Trifluoroacetate
Fournisseur: Bachem Americas
Certificats
A propos de cet article
Patented by Chromogenix AB, Sweden (CH 622 285). A highly specific substrate for human plasmin. For rat plasmin, Heinemann et al. determined a Km of 18.3 μM. Also cleaved by transmembrane tryptase (TMT).
Customers Who Bought This Also Bought