Ac-Gly-Pro-AFC
Fournisseur: Bachem Americas
Certificats
A propos de cet article
Ac-GP-AFC, specific substrate for the endopeptidase activity of fibroblast activation protein (FAP) (Km = 330 ± 30 μM; kcat = 7.7 ± 0.2 s⁻ ¹).
Customers Who Bought This Also Bought