Ac-Gly-Ala-Lys(Ac)-AMC
Fournisseur: Bachem Americas
Certificats
A propos de cet article
See also Boc-Lys(Ac)-AMC (I-1875) and Boc-Lys(Tfa)-AMC (I-1985).
Ac-GAK(Ac)-AMC, fluorogenic substrate for measuring histone deacetylase class I (HDAC 1, 2, 3, and 8) and class II (HDAC 6 and 10) activity in a protease-coupled assay. HDAC-catalyzed deacetylation of Lys yields Ac-GAK-AMC (I-1980).
Customers Who Bought This Also Bought