Z-Leu-Arg-Amc
Fournisseur: Bachem Americas
Certificats
A propos de cet article
Z-LR-AMC, a fluorogenic substrate for cathepsin K, L, S, and V, human tissue kallikrein, and the falcipains I - III. Z-LR-AMC is the preferred substrate of falcipain II, a hemoglobin-degrading cysteine protease from Plasmodium falciparum, one of the most promising targets for antimalarial drug design.
Customers Who Bought This Also Bought