Ac-Nle-Pro-Nle-Asp-AMC
Supplier: Bachem Americas
Certificates
About this item
Substrates for measuring the chymotryptic, PGPH, and tryptic acitivity of the 20S or 26S proteasome. See also the thrombin substrate I-1140.
Substrate for proteasome.
Customers Who Bought This Also Bought