Z-Val-Ala-Asn-AMC
Fournisseur: Bachem Americas
Certificats
A propos de cet article
Substrate for the determination of a new cysteine endopeptidase which is required for antigen processing. Z-VAN-AMC has been used as a fluorogenic substrate for porcine asparaginyl endopeptidase (AEP, legumain).
Customers Who Bought This Also Bought