Z-Arg-Leu-Arg-Gly-Gly-AMC Acetate
Fournisseur: Bachem Americas
Certificats
A propos de cet article
Z-RLRGG-AMC is a fluorogenic substrate for the deubiquitinating enzyme isopeptidase T (IPaseT) and other ubiquitin C-terminal hydrolases (UCHs) based on the C-termini of ubiquitin with a kcat/Km value of 95 M⁻¹s⁻¹. Together with our product I-1685 it is used in continuous assays for precise mechanistic studies or microtiter plate assays for high-throughput inhibitor screening.
Customers Who Bought This Also Bought