Ac-Tyr-Val-Ala-Asp-AMC
Fournisseur: Bachem Americas
Certificats
A propos de cet article
Ac-YVAD-AMC is a fluorogenic substrate for caspase-1 (ICE). It displayed a Km of 8.5 ± 0.5 µM for recombinant human caspase-1.
Customers Who Bought This Also Bought