H-Pro-Arg-AMC · 2 HCl
Catalog # I-1605.0050BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:546103-85-9
- Size:50 mg
- Protein/peptide name:H-Pro-Arg-AMC · 2 HCl
- MDL No.:MFCD00238138
- Sequence:H-Pro-Arg-AMC · 2 HCl
- Molecular weight:501.41
- Molecular formula:C₂₁H₂₈N₆O₄ · 2HCI
- Storage conditions:–20 ± 5 °C
- Cat. no.:I-1605.0050BA
- Supplier No.:I-1605.0050
Specifications
About this item
PR-AMC, a fluorogenic Plasmodium falciparum dipeptidyl aminopeptidase 1 (DPAP1, cathepsin C) substrate.
Customers Who Bought This Also Bought