MeOSuc-Ala-Ala-Pro-Val-AMC
Fournisseur: Bachem Americas
Certificats
A propos de cet article
See also C-1040, C-2345, I-1000, and L-1435.
A highly sensitive fluorogenic substrate for the assay of human leukocyte and porcine pancreatic elastases. MeOSuc-AAPV-AMC is also cleaved by myeloblastin.
Customers Who Bought This Also Bought