Spatulas, Miniature, Aluminum Handle, Excelta Corp®
Fournisseur: Excelta
Certificats
A propos de cet article
Placing epoxy/lubricants in very small amounts.
- Miniature design for accessibility under scope
- Color coded width sizes for identification
- Different tip widths to fit different applications
These mini-spatulas (oilers) all feature 2.5" long with aluminum handles. Mini-spatulas are ideal for placing epoxies or lubricants in very small places. They are all conductive. They are electrically conductive. The color coated handles identify the tip width.
Spatula shape: Flat end blade/straight handle.
Customers Who Bought This Also Bought