PCH Series Personal Benchtop Cooler/Heater, Grant Instruments
Fournisseur: GRANT USA INC.
Certificats
A propos de cet article
The PCH series of personal benchtop cooler/heaters are ideal for laboratory applications requiring cooling and heating in microtubes within the temperature setting range of –10 to 100°C (14 to 212°F)
- Cooling/heating setting range from -10°C to 100°C, with very rapid cool down and heat-up times
- Stability ±0.1°C
- Choice of three models: capacity for up to 32 microtubes in a combination of two sizes (PCH-1) or up to 20 microtubes of one size (PCH-2 and PCH-3)
- Convenient integral reaction timer with audible alarm
The PCH series offer a very effective tool for applications in DNA sample preparation for denaturing electrophoresis. The exceptionally rapid heat-up time enables the denaturation temperature of 95°C to be reached swiftly. The timer will signify denaturation time-up, and the samples can be rapidly cooled again and readied to load the gels.
The PCH series of cooler/heaters enable a stable temperature to be maintained during room temperature fluctuations.
Certifications: CE marked.
Ordering information: Units are supplied with cooling/heating block, and manufacturer's two-year warranty.
Spécifications
- Catalog No:
- 89402-936
- 97025-548
- 97025-550
- Cool-down time:
- 100 to –10 °C in 28 min, 25 to –10 °C in 21 min
- 100 to –10 °C in 28 min, 25 to –10 °C in 21 min
- 100 to –10 °C in 28 min, 25 to –10 °C in 21 min
- Temperature control range:
- Ambient –30 to 100°C
- Ambient –30 to 100°C
- Ambient –30 to 100°C
- Temperature resolution:
- 0.1°C
- 0.1°C
- 0.1°C
- Temperature stability:
- ±0.1°C
- ±0.1°C
- ±0.1°C
- Timer:
- 1 min. – 96 hrs. (Continuous)
- 1 min. – 96 hrs. (Continuous)
- 1 min. – 96 hrs. (Continuous)
- Display:
- 2 x 16 character LCD
- 2 x 16 character LCD
- 2 x 16 character LCD
- Weight:
- 3.6 kg (7.9 lbs.)
- 3.6 kg (7.9 lbs.)
- 3.6 kg (7.9 lbs.)