Distillation Receiver, Use At Other Than Atmospheric Pressure, Ace Glass
# de catalogue 89074-456
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Distillation Receiver
- Cat. no.:89074-456
- Supplier No.:9375-08
Spécifications
A propos de cet article
This receiver is designed for operation at other than atmospheric pressure
Vacuum is applied to the side arm with the T-bore stopcock. The secondary receiver is vented by means of the lower stopcock. The capacity from the bottom of the drip to the stopcock is 2.5 mL. The apparatus has a [ST] 14/20 outer joint at top, [ST] 14/20 inner joint at bottom. Hose connections are for use with 3/8-inch I.D. Both stopcocks are glass. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought