Apparel Dispensers, 1 Compartment, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
1 compartment dispenser with a closed front designed for cleanroom apparel.
Made of 1/4" (6mm) acrylic or PETG material.
Wall mounts or free-standing.
Wall mount bracket and support bar included.
Spécifications
- Catalog No:
- 14007-122
- 89208-656
- PPE type:
- Garments
- Garments
- Material:
- Acrylic
- PETG
- No. of compartments:
- 1
- 1
- Dispenser type:
- Floor-standing/wall-mountable
- Floor-standing/wall-mountable
- Thickness:
- 6 mm
- 6 mm