A propos de cet article
Compact digital block heaters with choice of models for 1 or 2 or 4 interchangeable blocks.
- Stability and uniformity ±0.1ºC
- Digitsl temperature control for optimum precision
- Heating range: ambient +5ºC to 130ºC, with rapid heat-up time
- Range of convenient features including alarms, single and dual point calibration, programmed start/stop, 'offset' for known sample temperature variation and choice of external or internal probes
- External probe available for accurate temperature control in a tube
Units feature an LED temperature display, easy to use interactive user interface for fast and accurate set up
The QB ranges are excellent all-round flexible block heaters where samples need heating, whether the application is general, industrial, clinical or life-science based.
The Grant QBD/QBH Digital Block Heaters features a heating range of ambient +5 to 130°C (all three QBD models) or ambient +5 to 200°C (QBH2 model). Units offer a rapid heat-up time, and temperature stability and uniformity of ± 0.1°C.
Variable high and low temperature alarms can be set to within 0.5 °C of the set point. Heaters are made from cool wall aluminium and chemically resistant plastic, have a low profile, a sloping fascia for ease of use, off-set adjustment for high accuracy operating temperatures and 0.1 °C display resolution.
Spécifications
- Catalog No:
- 97011-886
- 97011-890
- Heating time:
- 25 to 100°C in 15 mins.
- 25 to 100°C in 15 mins.
- Safety over-temperature cut-out:
- Thermal Fuse
- Thermal Fuse