Vessels, Freeze Drying, Wide Neck, Ace Glass
Supplier: Ace Glass
Certificates
About this item
These freeze drying vessels have 48 mm O.D
Working capacity of flasks is about one half the volume listed.
neck to fit #50 Ace-Thred bushing on 89055-792 connecting adapter for connection to the 89055-766 "umbrella" manifold. The 300 mL size has straight sides, with stops on each side to prevent “suck in” under vacuum; other sizes have sloped sides. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought