Filter Funnels, Hirsch, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
This Hirsch style glass filter funnel is useful when it is necessary to wash the precipitation and redissolve with chemicals that would attack filter paper normally used to for such a process
The angle of the funnel is 60°. Available in two different capacities and porosities. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought