Freeze Drying Vessels with Screw Cap, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
This freeze drying vessel has a GCMI-24-410 bottle thread at top and comes complete with a PTFE lined cap.
The neck also fits inside a #25 Ace-Thred bushing, so it can be used with a 89055-766 umbrella manifold. After drying the material, the vessel can be capped and stored.
Ordering information: For bushings, see part 89059-226. Working capacity of flasks is about one-half the volume listed.
Customers Who Bought This Also Bought