ContactUsLinkComponent
Vertical Mini Gel System, IBI Scientific
Vertical Mini Gel System, IBI Scientific
# de catalogue 89126-742
Fournisseur:  IBI Scientific
Vertical Mini Dual Gel System
Vertical Mini Gel System, IBI Scientific
# de catalogue 89126-742
Fournisseur:  IBI Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Max. sample capacity:
    15
  • Unit dimensions:
    18W x 17D x 17H cm
  • Description:
    MaGELin Cast-Your-Own Protein System
  • Cat. no.:
    89126-742
  • Gel Dimensions:
    10W x 10L cm
  • Supplier No.:
    IB94000
  • Gel dimensions:
    10 x 10 cm
  • Power supply:
    Max. 150 V
Spécifications
A propos de cet article
The MaGELin protein system is a versatile system designed to allow you to cast your own gels using the 10x10cm glass plates and easy-to-use casting fixture
The MaGELin IB94000 comes with three sets of glass plates, two sets of unique side sealing spacers, and a two place casting fixture for casting your own gels. The two place casting system has a very small footprint, 10x15cm, utilizing minimal space on your lab's bench-top. The gel sandwich is assembled by placing the glass plates into the matched grooves in the side spacers. Once assembled, the glass plate sandwiches are placed into the casting fixture base, as the lid is placed onto the base, the gel sandwiches are gently pressed down into the casting fixture gaskets and sealed. The gel can now be cast through the opening in the lid. Once the gel has polymerized, the glass plate sandwiches can be removed and placed into the gel capture device, and then into the buffer tank and run. The buffer tank holds a minimum of 350mL of buffer, and a maximum of 850mL for longer runs that may generate more heat. A stir bar can be placed into the buffer tank to circulate the buffer keeping the ionic concentration constant.
If you prefer to use precast gels the MaGELin protein system accommodates many of those as well. The MaGELin also utilizes a patented vented lid to help reduce heat buildup during a gel run.
Customers Who Bought This Also Bought