Spécifications
- Description:Data Acquisition Software
- Cat. no.:97011-798
- Supplier No.:840052
Spécifications
A propos de cet article
Software downloads data to your PC.
- Provides further analysis by receiving the data stream from your meter
- Data is saved in a text format (.txt) and can easily imported into spreadsheets, databases or word processing programs
Up to 16000 sample readings with real-time clock memory. Programmable sample interval from 1 second to 3,600 seconds. Unit displayed with changing setting. User-defined maximum (High) and minimum (Low) alarm setting. Includes one CD Rom and this operation manual.
System Requirements include Win 95 or higher operating system, 486 PC or higher withat least one serial port, RS232 Cable model 840054 sold separately above.
Works with meter models 800046, 840003, 840028, 840080, 840081, 840082, 840083, and 840085.