Environmental Quality Meter, Sper Scientific
# de catalogue 14223-290
Fournisseur: Sper Scientific
Spécifications
- Description:Environmental Quality Meter
- Cat. no.:14223-290
- Supplier No.:850071
Spécifications
A propos de cet article
Using interchangeable probes (sold separately) this versatile environmental meter reads airspeed, air temperature, RH, light and thermocouple and IR temperature
You can order probes for only the parameters currently required and add additional probes in the future. Intelligent probes electronically retain calibration information. No recalibration is required when changing probes. The display simultaneously reads temperature in user selectable ºC and ºF and all measurements are automatically temperature compensated. The RS232 port enables communication with a computer. The environmental quality meter also features automatic shut off, hold and min/max, has a fold out easel back and a tripod screw.
Ordering information: Comes with a 9V battery and a hard-shell foam-lined carrying case with room for one or two probes.
Customers Who Bought This Also Bought