A propos de cet article
This UV Light Meter is designed to provide measurements in a wide variety of applications
UV light intensity appears on the large backlit display in either µW or mW/cm², while the small lower display simultaneously shows minimum, maximum, average, and recorded data. Up to 20 data points can be recorded automatically at a variety of intervals. The unit features zero point reset, hold, and indicates when the battery is low. It is supplied in a protective hard foam-lined carrying case complete with a UV probe, tripod screw, magnetic mount, and 9V battery. Model 97036-014 measures short-wave UV-C light for applications such as UV light fume hoods, UV sterilization, industrial glass, and non-destructive testing. Model 97036-010 measures long- and medium-wave UV-A/B for applications such as forensics, chromatography, electrophoresis, and dermatology.
The Certified Light Meters (97036-012 and 97036-016) include a N.I.S.T. traceable certificate of calibration. Each unit is also supplied with a 5-year warranty.
It measures damaging UV (ultraviolet) light in the environment and museums. It provides UV degradation studies in the pharmaceutical, cosmetic, semiconductor, printing, and window tinting industries, among others. The unit may also be applied in laboratory chromatography, electrophoresis. and forensics. Finally, it can be used to test UV sterilization and dermatology lamps in hospitals.
Spécifications
- Catalog No:
- 97036-010
- 97036-014
- 97036-016
- Weight:
- 135 g (4.7 oz.)
- 135 g (4.7 oz.)
- 135 g (4.7 oz.)
- Dimensions:
- 145 x 55 x 40 mm (53/4 x 21/8 x 11/2")
- 145 x 55 x 40 mm (53/4 x 21/8 x 11/2")
- 145 x 55 x 40 mm (53/4 x 21/8 x 11/2")