Distillation Receivers, Separate Fractions, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
This distillation receiver is for use where separate fractions are desired without disturbing the distillation pressure
Available with [ST] 14/20 or [ST] 24/40 joints. Receiver flasks on [ST] 14/20 size are 5 mL; on [ST] 24/40 size, 50 mL. [ST] 14/20 size has hose connections for use with 3/8-inch or 5/16-inch I.D. tubing; [ST] 24/40 size has hose connections for use 3/8-inch I.D. tubing. Fabricated from borosilicate glass.
Ordering information: Complete [ST] 14/20 size includes 89074-458 vacuum adapter; 89074-460 distributor; four 89074-584 flasks (5 mL); and five 89059-608 Dupont Delrin® joint clips/clamps. Complete [ST] 24/40 size includes 89055-742 vacuum adapter; 89055-744 distributor; four 89056-176 flasks (50 mL); and five 89059-612 Dupont Delrin® joint clips/clamps. Individual components may also be purchased separately. For PTFE-clad, [ST] 14/20 size version, see 89074-462.
Customers Who Bought This Also Bought