Distillation Receiver, Separate Fractions, PTFE-Clad, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
This distillation receiver is for use where separate fractions are desired without disturbing the distillation pressure
All inner joints are PTFE-clad; outer joints are polished. All joints are [ST] 14/20. Receiver flasks are 5 mL capacity. Hose connections are for use with 3/8-inch or 5/16-inch I.D. tubing. Fabricated from borosilicate glass.
Ordering information: Complete item includes 89074-464 vacuum adapter; 89074-460 distributor; four 89074-596 flasks (5 mL); and five 89059-608 Dupont Delrin® joint clips/clamps. For non-PTFE-clad version, see 89074-476. For [ST] 24/40 non-PTFE-clad version see 89055-730.
Customers Who Bought This Also Bought