VWR® Stereo Zoom Binocular Microscope
Catalog # 89404-478
Supplier: VWR International
Specifications
- Description:VWR® Stereo Zoom Binocular Microscope
- Cat. no.:89404-478
Specifications
About this item
The VWR® Stereo Zoom Binocular Microscope is a high-performance unit designed for life science, industrial and pharmaceutical inspection, electronic assembly, and bonding operations.
- Binocular stereo microscope head
- Widefield eyepieces
- Transmitted illumination
The microscope features the Greenough Zoom Optical System, a binocular head with 360° rotation, photo/video: 100% [right eyetube], and widefield eyepieces 10X/F.N. 23. The unit offers a 6.7:1 zoom ratio, WD = 113mm, and a magnification range of 0.75X–5X. Addtional standard features include incident/transmitted large working area stand, 12V/10W intensity-controlled halogen incident, and transmitted illumination. Main power supply is 100V–240V.
Ordering information: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.
Customers Who Bought This Also Bought