ContactUsLinkComponent
VWR® Clinical Laboratory Trinocular Microscope
VWR® Clinical Laboratory Trinocular Microscope
# de catalogue 89404-474
Fournisseur:  VWR International
cUL Listed
CSA Certified
UL Listed
VWR* Collection Clinical Lab Trinocular
VWR® Clinical Laboratory Trinocular Microscope
# de catalogue 89404-474
Fournisseur:  VWR International
cUL Listed
CSA Certified
UL Listed
Spécifications
  • Description:
    VWR® Clinical Laboratory Trinocular Microscope
Spécifications
A propos de cet article
The VWR® Clinical Laboratory Microscope features a trinocular Siedentopf type head with a 30° inclination and 360° rotating capabilities.
The VWR® Clinical Laboratory Microscope features a trinocular Siedentopf type head with a 30° inclination and 360° rotating capabilities. Light distribution is 100/0, 20/80. Additional standard features include widefield eyepieces WF 10X/20mm, reversed quintuple nosepiece, planachromatic objectives PL 4X, phase contrast planachromatic objectives PLPh 10X, 40X S, 100X S-Oil, coaXial coarse and fine focusing adjustment, built-in low position coaXial mechanical stage, special five-hole turret type Abbe condenser 1.25 N.A. with choices for phase contrast 10X, 40X, 100X, brightfield and darkfield attaching, centering telescope, and Koehler illumination halogen lamp (12V/20W) with intensity control.
  • Ergonomic, anti-fatigue design
  • Widefield eyepieces
The VWR® Clinical Laboratory Microscope features a trinocular Siedentopf type head with a 30° inclination and 360° rotating capabilities.
Light distribution is 100/0, 20/80
Clinical Laboratory Trinocular Microscope features Planachromatic and Phase Contrast objectives that are ideal for research work. These features, combined with the ergonomic, anti-fatigue design allow for long hours of comfortable observation.
Additional standard features include widefield eyepieces WF 10X/20mm, reversed quintuple nosepiece, planachromatic objectives PL 4X, phase contrast planachromatic objectives PLPh 10X, 40X S, 100X S-Oil, coaxial coarse and fine focusing adjustment, built-in low position coaxial mechanical stage, special five-hole turret type Abbe condenser 1.25 N.A. with choices for phase contrast 10X, 40X, 100X, brightfield and darkfield attaching, centering telescope, and Koehler illumination halogen lamp (12V/20W) with intensity control.
Certifications: C-UL, UL Listed. CSA certified.
Ordering information: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.
Customers Who Bought This Also Bought