VWR® Stereo Basic Halogen 2x-4x Microscope
Fournisseur: VWR International
Certificats
A propos de cet article
The VWR® Stereo Basic Halogen 2X–4X Microscope is designed for use in electronics inspection, education, and life science applications.
The VWR® Stereo Basic Halogen 2X–4X Microscope features a 45° binocular stereo microscope head, widefield eyepieces 10X/F.N. 20, turret objective magnification of 2X–4X, 12V/10W intensity-controlled halogen incident, and transmitted illumination. Main power supply is 100V–240V.
- Binocular stereo microscope head
- Widefield eyepieces
- Transmitted illumination
The VWR® Stereo Basic Halogen 2X–4X Microscope features a 45° binocular stereo microscope head, widefield eyepieces 10X/F.N. 20, turret objective magnification of 2X–4X, 12V/10W intensity-controlled halogen incident, and transmitted illumination. Main power supply is 100V–240V.
Ordering information: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.
Customers Who Bought This Also Bought