Pocket Light Meter, Sper Scientific
# de catalogue 97035-924
Fournisseur: Sper Scientific
Spécifications
- Description:Pocket Light Meter, Certified
- Dimensions:114 x 76 x 19 mm (41/2 x 3 x 3/4")
- Cat. no.:97035-924
- Supplier No.:840010C
Spécifications
A propos de cet article
Easily transported in a shirt pocket, this tiny light meter is less than ¾” thick, and weighs only 120g (4.2oz.)
The Pocket Light Meter is certified, and includes a N.I.S.T. traceable certificate of calibration. The unit is also supplied with a 5-year warranty.
The controls, display, and sensor are all neatly contained and protected within the folding case with directions printed inside the cover. When open the sensor may be extended for remote use or snaps into a fixed position atop the case. Unit features a bar graph display, auto power off, and hold functions. It also indicates low battery and over range. Powered by 2 button cell batteries (included).
Customers Who Bought This Also Bought