Distillation Receivers, All-Purpose, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
This is an excellent all-purpose receiver for general distillation work
Equipped with a complete stopcock manifold to allow contents of the receiver to be removed to a single receiving flask without disturbing the vacuum of the system. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought